missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG12 Polyclonal Antibody

ATG12 Polyclonal Antibody
Brand: Invitrogen PA517218
This item is not returnable.
View return policy
Description
ATG12 Polyclonal Antibody for Western Blot, ICC/IF, IP
Specifications
| ATG12 | |
| Polyclonal | |
| Unconjugated | |
| ATG12 | |
| 4931423H11Rik; A330058M13Rik; APG 12; APG12; Apg12 (autophagy, yeast) homolog; APG12L; APG12-like; ATG12; ATG12 autophagy related 12 homolog; Atg12l; Atg12p; Atg12-PB; Autophagy protein 12-like; autophagy related 12; autophagy related protein 12; autophagy-related 12; autophagy-related protein 12; Autophagy-specific gene 12; CG10861; CG10861-PB; Dmel\CG10861; Dmel_CG10861; FBR93; HAPG12; IP05205p; IP05405p; OTTHUMP00000159129; ubiquitin-like protein ATG12; YBR1506; YBR217W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 67526 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 29 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q9CQY1 | |
| ATG12 | |
| Synthetic peptide corresponding to residues near the amino terminus of mouse Atg12. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction