missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG12 Monoclonal Antibody (GT166)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527801
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2, Neuro2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12 Predicted Reactivity: Pig (81%), Chimpanzee (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ATG12, a member of the autophagy protein family, forms a conjugate with ATG5; this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy. This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. ATG12 has also been shown to interact with ATG10 in human embryonic kidney cells in the presence of ATG7. At least two isoforms of ATG12 are known to exist. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1.
Specifications
| ATG12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O94817, Q9CQY1 | |
| ATG12 | |
| Recombinant protein encompassing a sequence within the center region of human ATG12. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot, Proximity Ligation Assay, Immunocytochemistry | |
| GT166 | |
| Unconjugated | |
| ATG12 | |
| 4931423H11Rik; A330058M13Rik; APG 12; APG12; Apg12 (autophagy, yeast) homolog; APG12L; APG12-like; ATG12; ATG12 autophagy related 12 homolog; Atg12l; Atg12p; Atg12-PB; Autophagy protein 12-like; autophagy related 12; autophagy related protein 12; autophagy-related 12; autophagy-related protein 12; Autophagy-specific gene 12; CG10861; CG10861-PB; Dmel\CG10861; Dmel_CG10861; FBR93; HAPG12; IP05205p; IP05405p; OTTHUMP00000159129; ubiquitin-like protein ATG12; YBR1506; YBR217W | |
| Mouse | |
| Protein G | |
| RUO | |
| 67526, 9140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction