missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG10 Rabbit anti-Human, Mouse, Clone: 2Q3Q3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16649-100UL
This item is not returnable.
View return policy
Description
ATG10 Monoclonal antibody specifically detects ATG10 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications
| ATG10 | |
| Monoclonal | |
| Unconjugated | |
| APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
| A synthetic peptide corresponding to a sequence within amino acids 121-220 of human ATG10 (Q9H0Y0). | |
| 100 μg | |
| Autophagy | |
| 83734 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| 2Q3Q3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction