missing translation for 'onlineSavingsMsg'
Learn More

ATG10 Antibody (SR1389), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen ATG10
Clone SR1389
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18654185
View Documents
Novus Biologicals
NBP3-21730-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18602085
View Documents
Novus Biologicals
NBP3-21730-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATG10 Monoclonal antibody specifically detects ATG10 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

ATG10
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human
APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10
A synthesized peptide derived from human ATG10 (Uniprot #: Q9H0Y0)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1389
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Autophagy
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
83734
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.