missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | ATG10 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ATG10 Polyclonal specifically detects ATG10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| ATG10 | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Autophagy | |
| PBS with 0.02% Sodium Azide | |
| APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10 | |
| ATG10 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9H0Y0 | |
| 83734 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human ATG10. The immunogen is located within the last 50 amino acids of ATG10. Amino Acid Squence: TPVLKNSQKINKNVN | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit