missing translation for 'onlineSavingsMsg'
Learn More

ATG10 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £359.00

Specifications

Antigen ATG10
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18682292
View Documents
Novus Biologicals
NBP2-92493-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18691232
View Documents
Novus Biologicals
NBP2-92493-0.1ml
0.1 mL
£359.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATG10 Polyclonal antibody specifically detects ATG10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ATG10
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Autophagy
PBS with 50% glycerol, pH7.3.
83734
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
APG10, APG10 autophagy 10-like (S. cerevisiae), APG10L, APG10-like, ATG10 autophagy related 10 homolog (S. cerevisiae), Autophagy-related protein 10, DKFZp586I0418, EC 6.3.2.-, FLJ13954, pp12616, ubiquitin-like-conjugating enzyme ATG10
A synthetic peptide corresponding to a sequence within amino acids 120-220 of human ATG10 (NP_113670.1). LPLSYAKATSQDERNVP
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.