missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38445-20ul
This item is not returnable.
View return policy
Description
ATF7 Polyclonal antibody specifically detects ATF7 in Human samples. It is validated for ELISA,Western Blot
Specifications
| ATF7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| activating transcription factor 7ATFATranscription factor ATF-A, cAMP-dependent transcription factor ATF-7, cyclic AMP-dependent transcription factor ATF-7, MGC57182 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human ATF7 (NP_001193611.1).,, Sequence:, MGDDRPFVCNAPGCGQRFTNEDHLAVHKHKHEMTLKFGPARTDSVIIADQTPTPTRFLKNCEEVGLFNELASSFEHEFKKAADEDEKKARSRTVAKKLVVFRPRLFLLCFGIIFLIG | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11016 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction