missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF7 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30071G
This item is not returnable.
View return policy
Description
ATF7 Polyclonal antibody specifically detects ATF7 in Human samples. It is validated for Western Blot
Specifications
| ATF7 | |
| Polyclonal | |
| Western Blot | |
| activating transcription factor 7ATFATranscription factor ATF-A, cAMP-dependent transcription factor ATF-7, cyclic AMP-dependent transcription factor ATF-7, MGC57182 | |
| maps to a region between residue 444 and 494 of human activating transcription factor 7 using the numbering given in entry CAA40483.1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 11016 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction