missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 beta Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84475-0.1ML
This item is not returnable.
View return policy
Description
ATF6 beta Polyclonal specifically detects ATF6 beta in Human samples. It is validated for Western Blot.
Specifications
| ATF6 beta | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1388 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| activating transcription factor 6 betacAMP-dependent transcription factor ATF-6 beta, ATF6-beta, cAMP response element-binding protein-related protein, cAMP responsive element binding protein-like 1, CREBL1CREB-RP, Creb-related protein, Creb-rp, cyclic AMP-dependent transcription factor ATF-6 beta, FLJ10066, G13cAMP-responsive element-binding protein-like 1, Protein G13 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human ATF6 beta | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction