missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATF6 Monoclonal antibody specifically detects ATF6 in Human, Mouse, Rat, Porcine, Plant, Rabbit samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
Specifications
| Antigen | ATF6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 70B1413.1 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry, Flow (Intracellular) |
| Formulation | 50mM Sodium Borate |
| Gene Alias | activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha |
| Host Species | Mouse |
| Immunogen | This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?