missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 Antibody (70B1413.1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-40256IR
This item is not returnable.
View return policy
Description
ATF6 Monoclonal specifically detects ATF6 in Human, Mouse, Rat, Porcine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated.
Specifications
| ATF6 | |
| Monoclonal | |
| DyLight 755 | |
| P18850 | |
| ATF6 | |
| This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. | |
| 0.1 mL | |
| Lipid and Metabolism, Signal Transduction, Unfolded Protein Response | |
| 22926 | |
| Human, Mouse, Rat, Pig, Rabbit | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay | |
| 70B1413.1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated | |
| activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This ATF6 antibody detects both the full length and the cleaved/active protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction