Learn More
ATF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10835-1-AP-20UL
 View more versions of this product
Product Code. 16877483
  • £135.00 / 20µL
Description
This antibody is a rabbit polyclonal antibody raised against full length human ATF4 antigen. The antibody recognizes the 38 kDa ATF4 protein and its phosphorylated forms (50 kDa). ATF4 can bind DNA as a homodimer and as a heterodimer. ATF4 is ubiquitinate
This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication.
Specifications
Specifications
ATF4
Polyclonal
Unconjugated
ATF4
ATF4, CREB 2, CREB2, DNA binding protein TAXREB67, TAXREB67, TXREB
Rabbit
Antigen Affinity Chromatography
RUO
11911, 468, 79255
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18848, Q06507, Q9ES19
ATF4
ATF4 Fusion Protein Ag1279
20 μL
Primary
Human, Rat, Mouse