missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATF3 Recombinant Rabbit Monoclonal Antibody (24GB6925)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556974
This item is not returnable.
View return policy
Description
ATF3 Recombinant Monoclonal Antibody for Western Blot
ATF3 (Activating transcription factor 3) is a member of the mammalian activation transcription factor/cAMP responsive element-binding (CREB) protein family of transcription factors. It encodes a protein with a calculated molecular mass of 22 kD. ATF3 represses rather than activates transcription from promoters with ATF binding elements. An alternatively spliced form of ATF3 (ATF3 delta Zip) encodes a truncated form ATF3 protein lacking the leucine zipper protein-dimerization motif and does not bind to DNA. In contrast to ATF3, ATF3 delta Zip stimulates transcription presumably by sequestering inhibitory co-factors away from the promoter. It is possible that alternative splicing of the ATF3 gene may be physiologically important in the regulation of target genes. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| ATF3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ATF3 | |
| activating transcription factor 3; ATF 3; ATF3; cAMP-dependent transcription factor ATF-3; cyclic AMP-dependent transcription factor ATF-3; Liver regeneration factor 1; Lrf1; LRF-1; LRFI; LRG-21; MGC137608 protein; transcription factor LRG-21; zgc:55526 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11910, 467 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 24GB6925 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18847, Q60765 | |
| ATF3 | |
| Recombinant protein of human ATF3 (aa 1-181). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction