Learn More
Invitrogen™ ATF3 Recombinant Rabbit Monoclonal Antibody (24GB6925)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556973
 View more versions of this product
Product Code. 30275102
  • £434.00 / 100µL
Description
ATF3 Recombinant Monoclonal Antibody for Western Blot
ATF3 (Activating transcription factor 3) is a member of the mammalian activation transcription factor/cAMP responsive element-binding (CREB) protein family of transcription factors. It encodes a protein with a calculated molecular mass of 22 kD. ATF3 represses rather than activates transcription from promoters with ATF binding elements. An alternatively spliced form of ATF3 (ATF3 delta Zip) encodes a truncated form ATF3 protein lacking the leucine zipper protein-dimerization motif and does not bind to DNA. In contrast to ATF3, ATF3 delta Zip stimulates transcription presumably by sequestering inhibitory co-factors away from the promoter. It is possible that alternative splicing of the ATF3 gene may be physiologically important in the regulation of target genes. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATF3
Recombinant Monoclonal
Unconjugated
ATF3
activating transcription factor 3; ATF 3; ATF3; cAMP-dependent transcription factor ATF-3; cyclic AMP-dependent transcription factor ATF-3; Liver regeneration factor 1; Lrf1; LRF-1; LRFI; LRG-21; MGC137608 protein; transcription factor LRG-21; zgc:55526
Rabbit
Affinity chromatography
RUO
11910, 467
-20°C
Liquid
Western Blot
24GB6925
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P18847, Q60765
ATF3
Recombinant protein of human ATF3 (aa 1-181).
100 μL
Primary
Human, Mouse