missing translation for 'onlineSavingsMsg'
Learn More

ATF3 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18427623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427623 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427623 Supplier Novus Biologicals Supplier No. NBP102935FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATF3 Polyclonal antibody specifically detects ATF3 in Human, Mouse, Rat, Mammalia samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias activating transcription factor 3cAMP-dependent transcription factor ATF-3, ATF3deltaZip2, ATF3deltaZip2c, ATF3deltaZip3, cyclic AMP-dependent transcription factor ATF-3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a region of human ATF3 (within residues 100-150). [UniProt# P18847]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 467
Target Species Human, Mouse, Rat, Mammalia
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.