missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11908-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can recognize all the isoforms of human ATF2. The molecular weight detected is larger than the theoretical value, which may be related to protein modification. About 65-68 kDa band detected in the articles.
The transcription factor ATF-2 (also called CRE-BP1) binds to both AP-1 and CRE DNA response elements and is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 interacts with a variety of viral oncoproteins and cellular tumor suppressors and is a target of the SAPK/JNK and p38 MAP kinase signaling pathways. Various forms of cellular stress, including genotoxic agents, inflammatory cytokines and UV irradiation, stimulate the transcriptional activity of ATF-2. Cellular stress activates ATF-2 by phosphorylation of Thr69 and Thr71. Both SAPK and p38 MAPK have been shown to phosphorylate ATF-2 at these sites in vitro and in cells transfected with ATF-2.Specifications
| ATF2 | |
| Polyclonal | |
| Unconjugated | |
| ATF2 | |
| ATF2, CRE BP1, CREB 2, CREB2, CREBP1, HB16, TREB7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11909, 1386, 81647 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15336, P16951, Q00969 | |
| Atf2 | |
| ATF2 Fusion Protein Ag2581 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction