missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF2 Antibody (PCRP-ATF2-1B4) - Azide and BSA Free, Novus Biologicals™
Description
ATF2 Monoclonal antibody specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Protein Array
Specifications
Specifications
| Antigen | ATF2 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Peptide Array |
| Classification | Monoclonal |
| Clone | PCRP-ATF2-1B4 |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug per 100-500 ug protein, Protein Array |
| Formulation | 10mM PBS |
| Gene Alias | activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?