missing translation for 'onlineSavingsMsg'
Learn More

ATF2 Antibody (PCRP-ATF2-1B4), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18360833 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360833 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18360833 Supplier Novus Biologicals Supplier No. NBP314001AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATF2 Monoclonal antibody specifically detects ATF2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF2
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-ATF2-1B4
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558
Host Species Mouse
Immunogen Recombinant full-length human ATF2 protein (Uniprot: P15336)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Immunology, Innate Immunity, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1386
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.