missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF2 Antibody (3D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26498-50ul
This item is not returnable.
View return policy
Description
ATF2 Monoclonal antibody specifically detects ATF2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| ATF2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 3D12 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
| A synthesized peptide derived from Human ATF2 [UniProt P15336] | |
| 50 μL | |
| Cancer, Chromatin Research, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators | |
| 1386 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction