missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Atenolol

Product Code. 15483819
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483819 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483819 Supplier Thermo Scientific Alfa Aesar Supplier No. J61199.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29122-68-7 | C14H22N2O3 | 266.341 g/mol

Cardioselective beta-adrenergic blocker. An antihypertensive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Cardioselective beta-adrenergic blocker
  • An antihypertensive
TRUSTED_SUSTAINABILITY

Specifications

CAS 29122-68-7
Melting Point 151°C to 155°C
Color White
Molecular Formula C14H22N2O3
MDL Number MFCD00057645
Quantity 1 g
Merck Index 14,859
Synonym atenolol, tenormin, tenormine, normiten, blokium, prenormine, tenoblock, atehexal, betablok, cuxanorm
InChI Key METKIMKYRPQLGS-UHFFFAOYSA-N
SMILES CC(C)NCC(COC1=CC=C(C=C1)CC(=O)N)O
IUPAC Name 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
Molecular Weight (g/mol) 266.341
PubChem CID 2249
ChEBI CHEBI:2904
Formula Weight 266.34
Physical Form Powder
Chemical Name or Material (+/-)-Atenolol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.