missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATE1 Recombinant Rabbit Monoclonal Antibody (24GB4720)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557344
This item is not returnable.
View return policy
Description
ATE1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Arginyl-tRNA-protein transferase (ATE1), also designated arginyltransferase 1, belongs to the R-transferase family of proteins. In order for a protein to be degraded via the ubiquitin pathway, arginylation of the protein is required. ATE1 plays an important role in this process, as it is important for the posttranslational conjugation of arginine to the N-terminal aspartate-, glutamate- and possibly cystine-contaning substrates. ATE1 is a 518 amino acid protein. Alternative splicing results in two distinct isoforms. ATE1, which is found as a monomer, can localize to the cytoplasm and/or the nucleus.
Specifications
| ATE1 | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95260, Q9Z2A5 | |
| ATE1 | |
| A synthesized peptide derived from human ATE1 (aa 39-52). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4720 | |
| Unconjugated | |
| ATE1 | |
| AI225793; arginine-tRNA-protein transferase 1; arginine-tRNA--protein transferase 1; arginyltransferase 1; arginyl-tRNA-protein transferase; arginyl-tRNA--protein transferase 1; ATE1; AW547406; R-transferase 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11101, 11907, 293526 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction