missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ATBF1/ZFHX3 Polyclonal specifically detects ATBF1/ZFHX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ATBF1/ZFHX3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q15911 |
| Gene Alias | Alpha-fetoprotein enhancer-binding protein, AT motif-binding factor, AT motif-binding factor 1, ATBF1alpha-fetoprotein enhancer binding protein, AT-binding transcription factor 1ATBT, ZFH-3, zinc finger homeobox 3, zinc finger homeobox protein 3, Zinc finger homeodomain protein 3, ZNF927 |
| Gene Symbols | ZFHX3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?