missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATBF1/ZFHX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34126
This item is not returnable.
View return policy
Description
ATBF1/ZFHX3 Polyclonal specifically detects ATBF1/ZFHX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ATBF1/ZFHX3 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q15911 | |
| ZFHX3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KAQVQVPQQSHQQILPQQQQNQLSIAQSHSALLQPSQHPEKKNKLVIKEKEKESQRERDSAEGGEGNTGPKETLPDA | |
| 0.1 mL | |
| Prostate Cancer | |
| 463 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Alpha-fetoprotein enhancer-binding protein, AT motif-binding factor, AT motif-binding factor 1, ATBF1alpha-fetoprotein enhancer binding protein, AT-binding transcription factor 1ATBT, ZFH-3, zinc finger homeobox 3, zinc finger homeobox protein 3, Zinc finger homeodomain protein 3, ZNF927 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction