missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soraprazan, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Description
Soraprazan (BYK61359) is a selective, reversible K-competitive inhibitor of the H,K-ATPase (Ki=6.4 nM), with an IC50 of 0.19 μM in gastric glands. Soraprazan binds to the H,K-ATPase with a Kd of 28.27 nM. Soraprazan shows immediate inhibition of acid secretion and is more than 2000-fold selective for H,K-ATPase over Na,K- and Ca-ATPases.
Specifications
Specifications
| Chemical Name or Material | Soraprazan |
| CAS | 261944-46-1 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H25N3O3 |
| Quantity | 1 mg |
| Synonym | BYK61359 |
| Solubility Information | DMSO : ≥ 150 mg/mL (408.23 mM) |
| SMILES | O[C@@H]1[C@@H](C2=CC=CC=C2)NC3=C(C=CN4C3=NC(C)=C4C)[C@H]1OCCOC |
| Molecular Weight (g/mol) | 367.44 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction