missing translation for 'onlineSavingsMsg'
Learn More

Atazanavir sulfate, MedChemExpress

Product Code. 16413327
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16413327

Brand: Medchem Express HY17367A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atazanavir (BMS-232632) sulfate, a highly selective HIV-1 protease inhibitor, is the first protease inhibitor approved for once-daily administration. Atazanavir sulfate is a substrate and inhibitor of CYP3A4, and an inhibitor and inducer of P-glycoprotein (P-gp). Atazanavir sulfate is also a SARS-CoV 3CLpro inhibitor with an IC50 of 3.49 μM.

Atazanavir (BMS-232632) sulfate, a highly selective HIV-1 protease inhibitor, is the first protease inhibitor approved for once-daily administration. Atazanavir sulfate is a substrate and inhibitor of CYP3A4, and an inhibitor and inducer of P-glycoprotein (P-gp). Atazanavir sulfate is also a SARS-CoV 3CLpro inhibitor with an IC50 of

Specifications

Chemical Name or Material Atazanavir sulfate
Physical Form Solid
CAS 229975-97-7
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C38H54N6O11S
For Use With (Application) COVID-19-anti-virus
Synonym BMS-232632 sulfate
Solubility Information DMSO : 166 mg/mL (206.74 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(OC)N[C@@H](C(C)(C)C)C(NN(CC1=CC=C(C2=NC=CC=C2)C=C1)C[C@H](O)[C@H](CC3=CC=CC=C3)NC([C@H](C(C)(C)C)NC(OC)=O)=O)=O.O=S(O)(O)=O
Molecular Weight (g/mol) 802.93
Formula Weight 802.93
Percent Purity 99.28%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.