missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin 7-Like 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99131-50ul
This item is not returnable.
View return policy
Description
Ataxin 7-Like 3 Polyclonal antibody specifically detects Ataxin 7-Like 3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Ataxin 7-Like 3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 4-8 uL/mg of lysate | |
| Ataxin-7-Like Protein 3, ATXN7L3, SAGA-Associated Factor 11 Homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Ataxin 7-Like 3. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 56970 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction