missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin-2-like protein Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | Ataxin-2-like protein |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Ataxin-2-like protein Polyclonal specifically detects Ataxin-2-like protein in Human samples. It is validated for Western Blot.Specifications
| Ataxin-2-like protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| A2DA2RP, A2LG, A2lp, ataxin 2 related protein, ataxin 2-like, Ataxin-2 domain protein, ataxin-2-like protein, Ataxin-2-related protein | |
| The immunogen is a synthetic peptide directed towards the middle region of human Ataxin-2-like protein (NP_009176.2). Peptide sequence ATKDKFTDSAIAMNSKVNGEHKEKVLQRWEGGDSNSDDYDLESDMSNGWD | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 11273 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title