Learn More
Ataxin-2 Antibody (05) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-26904JF669
Product Code. 30163863
  • £342.00 / 0.10mL
Description
Ataxin-2 Monoclonal antibody specifically detects Ataxin-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ataxin-2
Monoclonal
Janelia Fluor 669
ataxin 2, ataxin 2), ataxin-2, ATX2spinocerebellar ataxia 2 (olivopontocerebellar ataxia 2, autosomal dominant, SCA2FLJ46772, Spinocerebellar ataxia type 2 protein, TNRC13, trinucleotide repeat containing 13, Trinucleotide repeat-containing gene 13 protein
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Ataxin-2 (Uniprot#: Q99700; aa 733-942).
0.1 mL
Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers
6311
Store at 4°C in the dark.
IgG
ELISA
05
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified