missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin-10 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97393R
This item is not returnable.
View return policy
Description
Ataxin-10 Polyclonal specifically detects Ataxin-10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Ataxin-10 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| ataxin 10, ataxin-10, Brain protein E46 homolog, E46L, FLJ37990, HUMEEP, SCA10spinocerebellar ataxia 10, Spinocerebellar ataxia type 10 protein | |
| Produced in rabbits immunized with E. coli-derived Human Ataxin-10 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 25814 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction