Learn More
ATAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561340
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EVEPLCILDF YIHESVQRHG HGRELFQYML QKERVEPHQL AIDRPSQKLL KFLNKHYNLE TTVPQVNNFV IFE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
This gene encodes a protein that localizes to clathrin-coated pits, where it acetylates alpha tubulin on lysine 40. This process may be important in microtubule growth, for instance during chemotaxis and the formation of cilium. Alternative splicing results in multiple transcript variants.Specifications
ATAT1 | |
Polyclonal | |
Unconjugated | |
ATAT1 | |
0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255; alpha tubulin acetyltransferase 1; alpha-TAT; alpha-TAT {ECO:0000255; alpha-TAT1; alpha-TAT1 {ECO:0000255; alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255; alpha-tubulin N-acetyltransferase; alpha-tubulin N-acetyltransferase 1; Atat1; C23H6orf134; C6orf134; HAMAP-Rule:MF_03130}; hypothetical protein FLJ13158; MEC17; Nbla00487; Novel DUF738 containing protein (2610110G12Rik); RGD1303066; TAT; TAT {ECO:0000255 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79969 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.55 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SQI0 | |
ATAT1 | |
Recombinant protein corresponding to Human ATAT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only