missing translation for 'onlineSavingsMsg'
Learn More

ATAD4 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18331665 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331665 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331665 Supplier Novus Biologicals Supplier No. NBP306059R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATAD4 Polyclonal specifically detects ATAD4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATAD4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATAD4, ATPase family, AAA domain containing 4, MGC11242, proline rich 15-like, proline-rich protein 15-like protein, Protein ATAD4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ATAD4 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 79170
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.