Learn More
Invitrogen™ ATAD3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596988
Description
Immunogen sequence: MSWLFGVNKG PKGEGAGPPP PLPPAQPGAE GGGDRGLGDR PAPKDKWSNF DPTGLERAAK AARELEHSRY AKEALNLAQM QEQTLQLEQQ SKLKEYEAAV EQLKSEQIRA QAEERRKTLS EETRQHQARA QYQDKLARQR YEDQLKQQQL LNEENLRKQE ESVQKQEAMR RATVEREMEL RHKNEMLRVE TEARARAKAE RENADIIREQ IRLKASEHRQ TVLESIRTAG TLFGEGFRAF; Positive Samples: NIH/3T3, Mouse heart, Mouse spleen, Mouse brain, Rat brain; Cellular Location: Mitochondrion inner membrane, Peripheral membrane protein.
ATAD3B is a member of the AAA ATPase family, a family of proteins that catalyze ATP into ADP and are involved in several cellular functions such as cell-cycle regulation, protein proteolysis and transport. The AAA ATPase family is characterized by a highly conserved AAA motif containing Walker homology sequences and imparting ATPase activity. Mitochondrial membrane proteins ATAD3A/B contribute to the stabilization of nucleoids and may participate in the transformation pathway and the chemosensitivity of oligodendrogliomas. The gene encoding ATAD3A/B/C maps to human chromosome 1 and has been suggested to be an anti-apoptotic marker.
Specifications
| ATAD3B | |
| Polyclonal | |
| Unconjugated | |
| ATAD3B | |
| AAA-ATPase TOB3; AAA-TOB3; ATAD3B; ATPase family AAA domain-containing protein 3B; ATPase family, AAA domain containing 3B; KIAA1273; RP4-628J24.3; TOB3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83858 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5T9A4 | |
| ATAD3B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ATAD3B (NP_1141273). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.