missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55766
This item is not returnable.
View return policy
Description
ATAD3A Polyclonal specifically detects ATAD3A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ATAD3A | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| ATPase family AAA domain-containing protein 3A, ATPase family, AAA domain containing 3A, FLJ10709, FLJ35514 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| ATAD3A | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KLALHSGMDYAIMTGGDVAPMGREGVTAMHKLFDWANTSRR | |
| 100 μL | |
| Protein Phosphatase | |
| 55210 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction