missing translation for 'onlineSavingsMsg'
Få mere at vide

ATAD2 Antibody (SR1273), Novus Biologicals™

Produktkode 18606504 Shop alle Bio Techne produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
50 μg
Förpackningsstorlek
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
18606504 100 μg 100µL
18623744 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 18606504 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP322019100ul

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Rabbit Monoclonal Antibody

ATAD2 Monoclonal antibody specifically detects ATAD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ATAD2
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone SR1273
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias AAA nuclear coregulator cancer-associated protein, ANCCAMGC131938, ATPase family AAA domain-containing protein 2, ATPase family, AAA domain containing 2, DKFZp667N1320, EC 3.6.1.3, MGC142216, MGC29843, MGC5254, PRO2000
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATAD2 (Uniprot #: Q6PL18)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 29028
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.