missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AT7519 TFA, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
AT7519 (AT7519M) TFA as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively.
Specifications
Specifications
| Chemical Name or Material | AT7519 TFA |
| CAS | 1431697-85-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H18Cl2F3N5O4 |
| Quantity | 10 mg |
| Synonym | AT7519M TFA |
| Solubility Information | DMSO : 100 mg/mL (201.50 mM; Need ultrasonic) |
| SMILES | O=C(NC1=CNN=C1C(NC2CCNCC2)=O)C3=C(Cl)C=CC=C3Cl.O=C(O)C(F)(F)F |
| Molecular Weight (g/mol) | 496.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction