missing translation for 'onlineSavingsMsg'
Learn More

AT7519 TFA, MedChemExpress

Product Code. 16447906
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447906

Brand: Medchem Express HY50940A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AT7519 (AT7519M) TFA as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively.

AT7519 (AT7519M) TFA as a potent inhibitor of CDKs, with IC50s of 210, 47, 100, 13, 170, and <10 nM for CDK1, CDK2, CDK4 to CDK6, and CDK9, respectively.

Specifications

Chemical Name or Material AT7519 TFA
CAS 1431697-85-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H18Cl2F3N5O4
Quantity 100 mg
Synonym AT7519M TFA
Solubility Information DMSO : 100 mg/mL (201.50 mM; Need ultrasonic)
SMILES O=C(NC1=CNN=C1C(NC2CCNCC2)=O)C3=C(Cl)C=CC=C3Cl.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 496.27
Formula Weight 496.27
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.