missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AT 101
GREENER_CHOICE

Product Code. 18025082
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025082

Brand: Tocris Bioscience™ 3367/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Downregulates Bcl-2 and Mcl-1; pro-apoptotic

R-(-)-enantiomer of gossypol (Cat. No. 1964). Mimics the BH3 domains of Bcl-2, Bcl-XL and Mcl-1. Disrupts heterodimerization of Bcl-2 with proapoptotic family members. Induces apoptosis in vitro through activation of caspase-9; cytotoxic to multiple myeloma and drug-resistant cell lines. Delays onset of androgen-independent growth of VCaP prostate cancer xenografts in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90141-22-3
Molecular Formula C30H30O8

Specifications

Quantity 10 mg
Formula Weight 518.56
Purity >98%
Molecular Weight (g/mol) 518.562
Chemical Name or Material AT 101
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.