missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASZ1, Mouse anti-Human, Clone: 3C9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00136991-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: QILKCVELLLSRNADPNVACRRLMTPIMYAARDGHTQVVALLVAHGAEVNTQDENGYTALTWAARQGHKNIVLKLLELGANKMLQTKDGKMPSEIAKRNKHHEIFNLLSpecifications
| ASZ1 | |
| Monoclonal | |
| Unconjugated | |
| ankyrin repeat, SAM and basic leucine zipper domain containing 1 | |
| ALP1/ANKL1/C7orf7/GASZ/MGC26634/Orf3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3C9 | |
| In 1x PBS, pH 7.4 | |
| NM_130768 | |
| ASZ1 | |
| ASZ1 (NP_570124, 127 a.a. ∼ 234 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 136991 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction