missing translation for 'onlineSavingsMsg'
Learn More

ASZ1 Antibody (3C9), Novus Biologicals™

Product Code. 18414889 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414889 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414889 Supplier Novus Biologicals Supplier No. H00136991M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ASZ1 Monoclonal antibody specifically detects ASZ1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASZ1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3C9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_570124
Gene Alias ALP1Germ cell-specific ankyrin, SAM and basic leucine zipper domain-containingprotein, ANKL14933400N19Rik, ankyrin repeat, SAM and basic leucine zipper domain containing 1, ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1, ankyrin-like 1, Ankyrin-like protein 1, CT1.19, GASZC7orf7, MGC26634, Orf3
Host Species Mouse
Immunogen ASZ1 (NP_570124, 127 a.a. ∽ 234 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QILKCVELLLSRNADPNVACRRLMTPIMYAARDGHTQVVALLVAHGAEVNTQDENGYTALTWAARQGHKNIVLKLLELGANKMLQTKDGKMPSEIAKRNKHHEIFNLL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 136991
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.