missing translation for 'onlineSavingsMsg'
Learn More

ASXL2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18413643 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413643 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413643 Supplier Novus Biologicals Supplier No. NBP118926AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASXL2 Polyclonal antibody specifically detects ASXL2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASXL2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias additional sex combs like 2 (Drosophila), Additional sex combs-like protein 2, ASXH2polycomb group protein ASXH2, FLJ10898, KIAA1685DKFZp686C1968, putative Polycomb group protein ASXL2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 600 and 650 of human additional sex combs like 2 using the numbering given in entry NP_060733.4 (GeneID 55252).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55252
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.