missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASXL1 Monoclonal Antibody (11I4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA536238
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
ASXL1 Monoclonal antibody specifically detects ASXL1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot.
Specifications
| ASXL1 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4MG trehalose and 0.05MG sodium azide | |
| P59598, Q8IXJ9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 171023, 228790, 311553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 11I4 | |
| Unconjugated | |
| Asxl1 | |
| additional sex combs like 1; additional sex combs like 1, transcriptional regulator; additional sex combs like transcriptional regulator 1; additional sex combs-like protein 1; Asxl1; BOPS; KIAA0978; MDS; MGC117280; MGC71111; mKIAA0978; Polycomb group protein ASXL1; putative Polycomb group protein ASXL1; RGD1561878 | |
| A synthetic peptide corresponding to a sequence of human ASXL1 (KKERTWAEA ARLVLENYSDAPMTPKQILQVIEAE). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction