missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASXL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92590-0.1ml
This item is not returnable.
View return policy
Description
ASXL1 Polyclonal antibody specifically detects ASXL1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ASXL1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| additional sex combs like 1 (Drosophila) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-250 of human ASXL1 (NP_056153.2). LFYKLPGRISLFTLKKDALQWSRHPATVEGEEPEDTADVESCGSNEASTVSGENDVSLDETSSNASCSTESQSRPLSNPRDSYRASSQANKQKKKTGVMLPRVVLTPLKVNGAHVESASGFSGCHADGESGSPSSSSSGSLALGSAAIRGQAEVTQDPAPLLRGFRKPATGQMKRNRGEEI | |
| 0.1 mL | |
| Chromatin Modifiers, Signal Transduction | |
| 171023 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction