missing translation for 'onlineSavingsMsg'
Learn More

Asulam-d3 (methoxy-d3), CDN

Product Code. 30326657
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Product Code. Quantity unitSize
30326657 0.01 g 0.01g
30311446 0.005 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30326657

Brand: CDN CDND75310.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Asulam-d3 (methoxy-d3)
Molecular Formula C8 D3 H7 N2 O4 S
Quantity 0.01 g
Synonym Asulam-d3, Asulam-D3 (methoxy-D3), Methyl-d3 (4-aminophenylsulfonyl)carbamate, N1-(Methoxy-d3)carbonylsulfanilamide, N-[(4-Aminophenyl)sulfonyl]carbamic acid methyl-d3 ester
InChI Formula InChI=1 S/C8H10N2O4S/c1-14-8(11)10-15(12,13)7-4-2-6(9)3-5-7/h2-5 H,9H2,1H3,(H,10,11)/i1D3
SMILES [2 H]C([2 H])([2 H])OC(=O)NS(=O)(=O)c1ccc(N)cc1
IUPAC Name trideuteriomethyl N-(4-aminophenyl)sulfonylcarbamate
Molecular Weight (g/mol) 233.2595
Formula Weight 233.055 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.