missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Astrin Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74638CL1
This item is not returnable.
View return policy
Description
Astrin Polyclonal antibody specifically detects Astrin in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative),Immunocytochemistry/Immunofluorescence
Specifications
| Astrin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Astrin, Deepest, hMAP126, MAP126mitotic spindle coiled-coil related protein, mitotic spindle associated protein p126, Mitotic spindle-associated protein p126, sperm associated antigen 5, sperm-associated antigen 5 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human astrin (mitotic spindle associated protein p126) using the numbering given in entry NP_006452.3 (GeneID 10615). | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Stem Cell Markers | |
| 10615 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction