Learn More
Astrin Antibody (CL10078) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88942JF549
Product Code. 30298791
  • £558.00 / 0.10mL
Description
Astrin Monoclonal antibody specifically detects Astrin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Astrin
Monoclonal
Janelia Fluor 549
Astrin, Deepest, hMAP126, MAP126mitotic spindle coiled-coil related protein, mitotic spindle associated protein p126, Mitotic spindle-associated protein p126, sperm associated antigen 5, sperm-associated antigen 5
This antibody was developed using a synthetic peptide derived from Q96R06, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Stem Cell Markers
10615
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL10078
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified