missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Astrin Antibody (CL10078) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88942AF750
This item is not returnable.
View return policy
Description
Astrin Monoclonal antibody specifically detects Astrin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Astrin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Astrin, Deepest, hMAP126, MAP126mitotic spindle coiled-coil related protein, mitotic spindle associated protein p126, Mitotic spindle-associated protein p126, sperm associated antigen 5, sperm-associated antigen 5 | |
| This antibody was developed using a synthetic peptide derived from Q96R06, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Stem Cell Markers | |
| 10615 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL10078 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction