missing translation for 'onlineSavingsMsg'
Learn More

Astragaloside II, MedChemExpress

Product Code. 16498978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498978 10 mg 10mg
16441408 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498978 Supplier Medchem Express Supplier No. HYN0433
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Astragaloside II is a natural isolated from Astragalus.

Astragaloside II is a natural isolated from Astragalus.

Specifications

Chemical Name or Material Astragaloside II
CAS 84676-89-1
Color White
Purity Grade Notes Research
Molecular Formula C43H70O15
Quantity 10 mg
Synonym Astrasieversianin VIII
Solubility Information DMSO : 100 mg/mL (120.92 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](O)[C@@H](OC(C)=O)[C@]([H])(O[C@@H]2C(C)(C)[C@@]([C@@H](O[C@]3([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C[C@]4([H])[C@@]56CC[C@@]7(C)[C@@]4(C)C[C@H](O)[C@@]7([C@]8(C)O[C@H](C(O)(C)C)CC8)[H])([H])[C@]5(C6)CC2)OC1
Molecular Weight (g/mol) 827.01
Formula Weight 827.01
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.