missing translation for 'onlineSavingsMsg'
Learn More

Astragalin, MedChemExpress

Product Code. 16498818
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498818 10 mg 10mg
16451268 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498818 Supplier Medchem Express Supplier No. HYN0015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Astragalin (Astragaline) a flavonoid with anti-inflammatory, antioxidant, anticancer, bacteriostatic activity. Astragalin inhibits cancer cells proliferation and migration, induces apoptosis. Astragalin is orally active and provides nerve and heart protection, and resistance against and osteoporosis.

Astragalin (Astragaline) a flavonoid with anti-inflammatory, antioxidant, anticancer, bacteriostatic activity. Astragalin inhibits cancer cells proliferation and migration, induces apoptosis. Astragalin is orally active and provides nerve and heart protection, and resistance against and osteoporosis.

Specifications

Chemical Name or Material Astragalin
CAS 480-10-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20O11
Quantity 10 mg
Synonym Astragaline 3-Glucosylkaempferol Kaempferol 3-β-D-glucopyranoside
Solubility Information DMSO : ≥ 100 mg/mL (223.03 mM)
SMILES OC1=CC2=C(C(C(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C(C4=CC=C(O)C=C4)O2)=O)C(O)=C1
Molecular Weight (g/mol) 448.38
Formula Weight 448.38
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.