missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASTN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569433
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
This gene encodes a protein that is expressed in the brain and may function in neuronal migration, based on functional studies of the related astrotactin 1 gene in human and mouse. A deletion at this locus has been associated with schizophrenia. Multiple transcript variants encoding different proteins have been found for this locus.
Specifications
| ASTN2 | |
| Polyclonal | |
| Unconjugated | |
| ASTN2 | |
| 1d8; Astn2; Astnl; astrotactin 2; astrotactin 2-like; astrotactin-2; bA264C15.1; bA67K19.1; bM243L4.1 (astrotactin 2); bM264P11.1 (astrotactin 2); bM383P24.1 (astrotactin 2); bM423P8.1 (astrotactin 2); bM452J22.1; bM452J22.1 (novel protein); bM468K2.1 (astrotactin 2); Kiaa0634 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23245 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75129 | |
| ASTN2 | |
| synthetic peptide directed towards the N terminal region of human ASTN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction