missing translation for 'onlineSavingsMsg'
Learn More

ASTM Method D6550 4% Olefins Solution, Dr. Ehrenstorfer

Product Code. 30308829
Change view
Click to view available options
Quantity:
10 x 2 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30308829 10 x 2 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308829 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000874IE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Decene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣2,2,4-Trimethylpentane∣2-Methyl-1-butene∣2-Methylpentene-2∣3-Methyl-1-butene∣4-Methyl-1-pentene∣cis-2-Heptene∣cis-2-Hexene∣cis-2-Nonene∣cis-2-Octene∣cis-2-Pentene∣cis-3-Heptene∣cis-3-Nonene∣Ethanol∣Isoprene∣m-Xylene (1,3-Dimethylbenzene)∣Toluene∣trans-2-Heptene∣trans-2-Hexene∣trans-2-Nonene∣trans-2-Octene∣trans-2-Pentene∣trans-3-Heptene∣trans-3-Nonene
CAS 872-05-9, 592-76-7, 592-41-6, 124-11-8, 111-66-0, 109-67-1
Analyte or Component Names 1-Decene∣1-Heptene∣1-Hexene∣1-Nonene∣1-Octene∣1-Pentene∣2,2,4-Trimethylpentane∣2-Methyl-1-butene∣2-Methylpentene-2∣3-Methyl-1-butene∣4-Methyl-1-pentene∣cis-2-Heptene∣cis-2-Hexene∣cis-2-Nonene∣cis-2-Octene∣cis-2-Pentene∣cis-3-Heptene∣cis-3-Nonene∣Ethanol∣Isoprene∣m-Xylene (1,3-Dimethylbenzene)∣Toluene∣trans-2-Heptene∣trans-2-Hexene∣trans-2-Nonene∣trans-2-Octene∣trans-2-Pentene∣trans-3-Heptene∣trans-3-Nonene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Isooctane/Xylene/Ethanol
Quantity 10 x 2 mL
Concentration or Composition (by Analyte or Components) trans-2-Nonene 0.04048 %∣2-Methyl-1-butene 0.05864 %∣trans-2-Hexene 0.07104 %∣trans-2-Pentene 0.07396 %∣trans-3-Nonene 0.0752 %∣trans-2-Octene 0.08012 %∣3-Methyl-1-butene 0.0806 %∣cis-2-Pentene 0.08092 %∣Isoprene 0.0988 %∣cis-2-Nonene 0.11104 %∣2-Methylpentene-2 0.13608 %∣4-Methyl-1-pentene 0.13912 %∣trans-3-Heptene 0.149 %∣trans-2-Heptene 0.1508 %∣cis-2-Hexene 0.15848 %∣cis-2-Octene 0.15928 %∣cis-3-Nonene 0.16296 %∣1-Pentene 0.16856 %∣cis-2-Heptene 0.23284 %∣cis-3-Heptene 0.23672 %∣1-Decene 0.29392 %∣1-Heptene 0.30804 %∣1-Hexene 0.31012 %∣1-Nonene 0.31112 %∣1-Octene 0.31216 %∣m-Xylene (1,3-Dimethylbenzene) 4.8 %∣2,2,4-Trimethylpentane 9.6 %∣Ethanol 9.6 %∣Toluene 72 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.